3-Amino-N,N-dimethylbenzenesulfonamide 97%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-N,N-Dimethyl-Benzenesulphonamide

A182335-10G A182335-1G A182335-5G
77274-112EA 329.6 USD
77274-112 77274-114 77274-116
3-Amino-N,N-dimethylbenzenesulfonamide 97%
3-Amino-N,N-dimethylbenzenesulphonamide

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₂S
MW: 200.2600 g/mol
MDL Number: MFCD00602451
CAS Number: 6274-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR