Methanesulfonato(2-di-t-butylphosphino-3,4,5,6-tetramethyl-2',4',6'-tri-i-propylbiphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II) 98%

Supplier: Ambeed
Warning

Synonyms: 4MetBuXPhos Pd G3

A182143-100MG A182143-1G A182143-250MG A182143-5G
77248-304EA 19 USD
77248-304 77248-306 77615-540 77746-184
Methanesulfonato(2-di-t-butylphosphino-3,4,5,6-tetramethyl-2',4',6'-tri-i-propylbiphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II) 98%
Methanesulfonato(2-di-t-butylphosphino-3,4,5,6-tetramethyl-2',4',6'-tri-i-propylbiphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II)

Precaución: for Research Use Only

Formula: C₄₆H₆₆NO₃PPdS
MW: 850.48 g/mol
MDL Number: MFCD28009172
CAS Number: 1507403-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR