Ethyl 1-benzylpiperidine-3-carboxylate 98%

Supplier: Ambeed
Warning

A181999-10G A181999-5G
77274-108EA 13 USD
77274-108 77274-110
Ethyl 1-benzylpiperidine-3-carboxylate 98%
Ethyl 1-benzylpiperidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₂
MDL Number: MFCD00191221
CAS Number: 72551-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR