2,3,5,6-Tetrafluoronitrobenzene 97%

Supplier: Ambeed
Warning

Synonyms: 1,2,4,5-Tetrafluoro-3-nitrobenzene

A181955-10G A181955-250MG A181955-5G A181955-1G
77248-298EA 248.23 USD
77248-298 77248-300 77248-302 77746-180
2,3,5,6-Tetrafluoronitrobenzene 97%
2,3,5,6-Tetrafluoronitrobenzene

Precaución: for Research Use Only

Formula: C₆HF₄NO₂
MW: 195.071 g/mol
MDL Number: MFCD02093333
CAS Number: 6257-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR