4,5-Bis(hydroxymethyl)-2-phenylimidazole 96%

Supplier: Ambeed
Warning

Synonyms: (2-Phenyl-1H-imidazole-4,5-diyl)dimethanol

A181950-100MG A181950-1G A181950-250MG A181950-5G
77287-262EA 28.84 USD
77287-262 77287-264 77287-266 77287-268
4,5-Bis(hydroxymethyl)-2-phenylimidazole 96%
4,5-Bis(hydroxymethyl)-2-phenylimidazole

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O₂
MW: 204.23 g/mol
MDL Number: MFCD00047017
CAS Number: 61698-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR