4,7-Dimethoxy-1-indanone 98%

Supplier: Ambeed
Warning

A181842-100MG A181842-1G A181842-250MG A181842-5G
77287-254EA 5 USD
77287-254 77287-256 77287-258 77287-260
4,7-Dimethoxy-1-indanone 98%
4,7-Dimethoxy-1-indanone

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₃
MDL Number: MFCD00757244
CAS Number: 52428-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR