1-Boc-4-(6-chloropyridazin-3-yl)piperazine 98%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-4-(6-Chloropyridazin-3-yl)piperazine

A181819-1G A181819-250MG
77287-250EA 25.75 USD
77287-250 77287-252
1-Boc-4-(6-chloropyridazin-3-yl)piperazine 98%
1-Boc-4-(6-chloropyridazin-3-yl)piperazine

Precaución: for Research Use Only

Formula: C₁₃H₁₉ClN₄O₂
MW: 298.7700 g/mol
MDL Number: MFCD05864738
CAS Number: 492431-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR