2-Cyano-3-cyclopropylacrylic acid 97%

Supplier: Ambeed
Warning

A181759-100MG A181759-1G A181759-250MG
77236-586EA 146 USD
77236-586 77236-588 77236-590
2-Cyano-3-cyclopropylacrylic acid 97%
2-Cyano-3-cyclopropylacrylic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₂
MDL Number: MFCD22377851
CAS Number: 98895-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR