2-Amino-3-(ethylamino)pyrazine 95+%

Supplier: Ambeed
Warning

A181636-100MG A181636-1G A181636-250MG
77248-292EA 41.2 USD
77248-292 77248-294 77248-296
2-Amino-3-(ethylamino)pyrazine 95+%
2-Amino-3-(ethylamino)pyrazine

Precaución: for Research Use Only

Formula: C₆H₁₀N₄
MW: 138.1700 g/mol
MDL Number: MFCD08460071
CAS Number: 912773-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR