5-Bromo-2,3-dihydro-1H-indene-1-carbonitrile 97%

Supplier: Ambeed
Warning

A181564-1G A181564-250MG
77260-270EA 350.2 USD
77260-270 77260-272
5-Bromo-2,3-dihydro-1H-indene-1-carbonitrile 97%
5-Bromo-2,3-dihydro-1H-indene-1-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₈BrN
MW: 222.08 g/mol
MDL Number: MFCD11219692
CAS Number: 1188174-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR