2-Methyl-4-nitroindole 97%

Supplier: Ambeed
Warning

A181550-100MG A181550-250MG A181550-1G A181550-5G
77274-102EA 16.48 USD
77274-102 77274-104 77748-185 77748-186
2-Methyl-4-nitroindole 97%
2-Methyl-4-nitroindole

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.1700 g/mol
MDL Number: MFCD09028445
CAS Number: 3484-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR