H-Ser(tBu)-OBzl.HCl 97%

Supplier: Ambeed
Warning

Synonyms: H-Ser(tBu)-OBzl·HCl

A181517-1G A181517-250MG
77236-582EA 19.57 USD
77236-582 77236-584
H-Ser(tBu)-OBzl.HCl 97%
H-Ser(tBu)-OBzl.HCl

Precaución: for Research Use Only

Formula: C₁₄H₂₂ClNO₃
MW: 287.78 g/mol
MDL Number: MFCD20257234
CAS Number: 1401668-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR