(2,6-Dichlorophenyl)acetic acid 98%

Supplier: Ambeed
Warning

A181470-10G A181470-25G A181470-500G A181470-5G A181470-100G
77236-574EA 11 USD
77236-574 77236-576 77236-578 77236-580 77615-528
(2,6-Dichlorophenyl)acetic acid 98%
(2,6-Dichlorophenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₃Cl₂)CH₂COOH
MDL Number: MFCD00004320
CAS Number: 6575-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR