Pyridine-2-carbohydrazide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Picolinic acid hydrazide, 2-Pyridinecarbohydrazide, 2-Pyridinecarboxylic acid hydrazide, Picolinohydrazide, 2-Picolinyl hydrazide
A181361-100G
A181361-100MG
A181361-10G
A181361-1G
A181361-250MG
A181361-25G
A181361-5G
77274-088EA
364.62
USD
77274-088
77274-090
77274-092
77274-094
77274-096
77274-098
77274-100
Pyridine-2-carbohydrazide 98%
Pyridine-2-carbohydrazide
Precaución: for Research Use Only
Formula:
C₆H₇N₃O MW: 137.1393 g/mol |
MDL Number:
MFCD00059782 CAS Number: 1452-63-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...