N-(2,5-Difluoro-4-((2-(1-methyl-1H-pyrazol-4-yl)pyridin-4-yl)oxy)phenyl)-N-phenylcyclopropane-1,1-dicarboxamide 98+%

Supplier: Ambeed
Warning

Synonyms: c-Kit-IN-1

A181353-10MG A181353-1MG A181353-25MG A181353-50MG A181353-5MG
77236-564EA 38 USD
77236-564 77236-566 77236-568 77236-570 77236-572
N-(2,5-Difluoro-4-((2-(1-methyl-1H-pyrazol-4-yl)pyridin-4-yl)oxy)phenyl)-N-phenylcyclopropane-1,1-dicarboxamide 98+%
N-(2,5-Difluoro-4-((2-(1-methyl-1H-pyrazol-4-yl)pyridin-4-yl)oxy)phenyl)-N-phenylcyclopropane-1,1-dicarboxamide

Precaución: for Research Use Only

Formula: C₂₆H₂₁F₂N₅O₃
MW: 489.47 g/mol
MDL Number: MFCD22571733
CAS Number: 1225278-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR