(R)-N-(1,3-Dimethyl-6-(2-methylpiperazin-1-yl)-2-oxo-2,3-dihydro-1H-benzo[d]imidazol-5-yl)-2-methoxybenzamide 98%

Supplier: Ambeed
Warning

Synonyms: GSK6853

A181180-10MG A181180-1MG A181180-25MG A181180-50MG A181180-5MG
77260-242EA 87.55 USD
77260-242 77260-244 77260-246 77260-248 77260-250
(R)-N-(1,3-Dimethyl-6-(2-methylpiperazin-1-yl)-2-oxo-2,3-dihydro-1H-benzo[d]imidazol-5-yl)-2-methoxybenzamide 98%
(R)-N-(1,3-Dimethyl-6-(2-methylpiperazin-1-yl)-2-oxo-2,3-dihydro-1H-benzo[d]imidazol-5-yl)-2-methoxybenzamide

Precaución: for Research Use Only

Formula: C₂₂H₂₇N₅O₃
MW: 409.48 g/mol
MDL Number: MFCD30489734
CAS Number: 1910124-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR