2-Methyl-3-(trifluoromethyl)benzyl bromide 98%

Supplier: Ambeed
Danger

Synonyms: 1-(Bromomethyl)-2-methyl-3-(trifluoromethyl)benzene

A181124-1G A181124-250MG A181124-5G
77274-078EA 38.11 USD
77274-078 77274-080 77274-082
2-Methyl-3-(trifluoromethyl)benzyl bromide 98%
2-Methyl-3-(trifluoromethyl)benzyl bromide

Precaución: for Research Use Only

Formula: C₉H₈BrF₃
MW: 253.06 g/mol
MDL Number: MFCD01631608
CAS Number: 261952-16-3
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR