2-((1R,5S,6S)-6-(Aminomethyl)-3-ethylbicyclo[3.2.0]hept-3-en-6-yl)acetic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: Mirogabalin

A181027-10MG A181027-1MG A181027-5MG A181027-250MG A181027-50MG
77260-236EA 61.8 USD
77260-236 77260-238 77260-240 77615-522 77748-184
2-((1R,5S,6S)-6-(Aminomethyl)-3-ethylbicyclo[3.2.0]hept-3-en-6-yl)acetic acid ≥98%
2-((1R,5S,6S)-6-(Aminomethyl)-3-ethylbicyclo[3.2.0]hept-3-en-6-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₂
MW: 209.28 g/mol
MDL Number: MFCD28411425
CAS Number: 1138245-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR