Ethyl 4-oxo-3-(propionyloxy)-4H-pyran-2-carboxylate 97%

Supplier: Ambeed
Warning

A180998-10G A180998-25G A180998-5G
77260-226EA 9.27 USD
77260-226 77260-228 77260-230
Ethyl 4-oxo-3-(propionyloxy)-4H-pyran-2-carboxylate 97%
Ethyl 4-oxo-3-(propionyloxy)-4H-pyran-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₆
MW: 240.20 g/mol
MDL Number: MFCD30607292
CAS Number: 2061980-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR