4-Bromo-2,6-dimethylanisole 97%

Supplier: Ambeed
Warning

A180953-100G A180953-10G A180953-1G A180953-25G A180953-500G A180953-5G
77248-268EA 68 USD
77248-268 77248-270 77248-272 77248-274 77248-276 77248-278
4-Bromo-2,6-dimethylanisole 97%
4-Bromo-2,6-dimethylanisole

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
MDL Number: MFCD00143259
CAS Number: 14804-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR