2-Bromo-5-chloroanisole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-4-chloro-2-methoxybenzene

A180787-100G A180787-10G A180787-1G A180787-25G A180787-500G A180787-5G
77260-214EA 125.66 USD
77260-214 77260-216 77260-218 77260-220 77260-222 77260-224
2-Bromo-5-chloroanisole 98%
2-Bromo-5-chloroanisole

Precaución: for Research Use Only

Formula: C₇H₆BrClO
MW: 221.48 g/mol
MDL Number: MFCD03790889
CAS Number: 174913-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR