3-Methylpyrrolidin-2-one 97%

Supplier: Ambeed
Warning

A180761-1G A180761-5G
77274-064EA 152 USD
77274-064 77274-066
3-Methylpyrrolidin-2-one 97%
3-Methylpyrrolidin-2-one

Precaución: for Research Use Only

Formula: C₅H₉NO
MDL Number: MFCD00005271
CAS Number: 2555-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR