2-Chloro-5-(trifluoromethyl)benzoyl chloride 97%

Supplier: Ambeed
Danger

A180664-1G A180664-25G A180664-5G
77260-208EA 20 USD
77260-208 77260-210 77260-212
2-Chloro-5-(trifluoromethyl)benzoyl chloride 97%
2-Chloro-5-(trifluoromethyl)benzoyl chloride

Precaución: for Research Use Only

Formula: C₈H₃Cl₂F₃O
MDL Number: MFCD01631634
CAS Number: 657-05-6
UN: 1760
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR