Bicyclo[3.2.1]octane-2,4-dione 97%
Supplier: Ambeed
|
Warning
|
A180597-250MG
A180597-100MG
77260-204EA
78.28
USD
77260-204
77748-182
Bicyclo[3.2.1]octane-2,4-dione 97%
Bicyclo[3.2.1]octane-2,4-dione
Precaución: for Research Use Only
Formula:
C₈H₁₀O₂ MW: 138.16 g/mol |
MDL Number:
MFCD24504246 CAS Number: 74896-14-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...