(R)-3-Hydroxymethylmorpholine 95%

Supplier: Ambeed
Warning

A180540-100MG A180540-1G A180540-250MG A180540-5G
77274-042EA 42.23 USD
77274-042 77274-044 77274-046 77274-048
(R)-3-Hydroxymethylmorpholine 95%
(R)-3-Hydroxymethylmorpholine

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂
MW: 117.1500 g/mol
MDL Number: MFCD06799479
CAS Number: 211053-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR