4-(trans-4-Ethylcyclohexyl)phenol 95%

Supplier: Ambeed
Warning

A180491-10G A180491-1G A180491-5G A180491-100G A180491-25G
77287-186EA 26.78 USD
77287-186 77287-188 77287-190 77748-180 77748-181
4-(trans-4-Ethylcyclohexyl)phenol 95%
4-(trans-4-Ethylcyclohexyl)phenol

Precaución: for Research Use Only

Formula: C₁₄H₂₀O
MW: 204.31 g/mol
MDL Number: MFCD06658179
CAS Number: 89100-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR