Sodium ((1,4-phenylenebis(methanylylidene))bis(7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl-3-ylidene))dimethanesulfonate 97%

Supplier: Ambeed
Warning

A180412-1G A180412-250MG
77248-258EA 12.36 USD
77248-258 77248-260
Sodium ((1,4-phenylenebis(methanylylidene))bis(7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl-3-ylidene))dimethanesulfonate 97%
Sodium ((1,4-phenylenebis(methanylylidene))bis(7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl-3-ylidene))dimethanesulfonate

Precaución: for Research Use Only

Formula: C₂₈H₃₂Na₂O₈S₂
MW: 606.65 g/mol
MDL Number: MFCD30573899
CAS Number: 90458-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR