4-Boc-1-(5-bromo-2-pyridyl)piperazine 96%
Supplier: Ambeed
|
Danger
|
Synonyms:
5-Bromo-2-(4-tert-butoxycarbonylpiperazin-1-yl)pyridine, tert-Butyl 4-(5-bromopyridin-2-yl)piperazine-1-carboxylate
A180382-10G
A180382-1G
A180382-5G
77287-174EA
65.92
USD
77287-174
77287-176
77287-178
4-Boc-1-(5-bromo-2-pyridyl)piperazine 96%
4-Boc-1-(5-bromo-2-pyridyl)piperazine
Precaución: for Research Use Only
Formula:
C₁₄H₂₀BrN₃O₂ MW: 342.237 g/mol |
MDL Number:
MFCD07369772 CAS Number: 153747-97-8 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...