2-Pentyl-1,3-propanediol 97%

Supplier: Ambeed
Warning

A180320-100G A180320-25G A180320-5G
77274-028EA 232 USD
77274-028 77274-030 77274-032
2-Pentyl-1,3-propanediol 97%
2-Pentyl-1,3-propanediol

Precaución: for Research Use Only

Formula: C₈H₁₈O₂
MDL Number: MFCD00801123
CAS Number: 25462-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR