(R)-(-)-3-Aminopyrrolidine dihydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (R)-Pyrrolidin-3-amine dihydrochloride

A180311-1G A180311-250MG A180311-25G A180311-5G
77287-166EA 12.36 USD
77287-166 77287-168 77287-170 77287-172
(R)-(-)-3-Aminopyrrolidine dihydrochloride 97%
(R)-(-)-3-Aminopyrrolidine dihydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₂Cl₂N₂
MW: 159.06 g/mol
MDL Number: MFCD00070604
CAS Number: 116183-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR