Ethyl-2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate

A180269-100G A180269-10G A180269-1G A180269-25G A180269-5G
77236-546EA 497.49 USD
77236-546 77236-548 77236-550 77236-552 77615-518
Ethyl-2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate 97%
Ethyl-2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₈H₆ClF₃N₂O₂
MW: 254.6 g/mol
MDL Number: MFCD01312372
CAS Number: 187035-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR