Ethyl 2-chlorothieno[3,2-d]pyrimidine-4-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A180267-100MG
A180267-1G
A180267-250MG
77236-540EA
84.46
USD
77236-540
77236-542
77236-544
Ethyl 2-chlorothieno[3,2-d]pyrimidine-4-carboxylate 97%
Ethyl 2-chlorothieno[3,2-d]pyrimidine-4-carboxylate
Precaución: for Research Use Only
Formula:
C₉H₇ClN₂O₂S MW: 242.68 g/mol |
MDL Number:
MFCD17214222 CAS Number: 1135299-23-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...