Sodium naphthalene-1-sulfinate 97%

Supplier: Ambeed
Warning

A180177-100MG A180177-250MG
77248-250EA 46 USD
77248-250 77248-252
Sodium naphthalene-1-sulfinate 97%
Sodium naphthalene-1-sulfinate

Precaución: for Research Use Only

Formula: C₁₀H₇NaO₂S
MDL Number: MFCD24113320
CAS Number: 64326-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR