3-Chloro-p-cresol 97%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-methylphenol, 1-Chloro-3-hydroxy-6-methylbenzene

A180082-10G A180082-1G A180082-25G A180082-500G A180082-5G
77274-014EA 23.69 USD
77274-014 77274-016 77274-018 77274-020 77274-022
3-Chloro-p-cresol 97%
3-Chloro-p-cresol

Precaución: for Research Use Only

Formula: C₇H₇ClO
MW: 142.58 g/mol
MDL Number: MFCD00060319
CAS Number: 615-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR