Quinoxaline-2,3-dione 98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dihydroxyquinoxaline, Quinoxaline-2,3(1H,4H)-dione

A180048-100G A180048-10G A180048-25G A180048-5G A180048-500G
77248-244EA 85.49 USD
77248-244 77248-246 77248-248 77615-516 77746-173
Quinoxaline-2,3-dione 98%
Quinoxaline-2,3-dione

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
MDL Number: MFCD00006723
CAS Number: 15804-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR