Toluene-4-sulfonamide 98%

Supplier: Ambeed
Warning

A180022-100G A180022-500G A180022-1000G
77287-158EA 6 USD
77287-158 77287-160 77593-144
Toluene-4-sulfonamide 98%
Toluene-4-sulphonamide

Precaución: for Research Use Only

Formula: C₇H₉NO₂S
MDL Number: MFCD00011692
CAS Number: 70-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR