4-Benzyloxy-3-chlorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Benzyloxy-3-chlorobenzeneboronic acid, (4-(Benzyloxy)-3-chlorophenyl)boronic acid

A179977-1G A179977-25G A179977-5G
77274-008EA 11.33 USD
77274-008 77274-010 77274-012
4-Benzyloxy-3-chlorophenylboronic acid 98%
4-Benzyloxy-3-chlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂BClO₃
MW: 262.50 g/mol
MDL Number: MFCD04115642
CAS Number: 845551-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR