2-Amino-5-bromonicotinic acid 97% (by HPLC)

Supplier: Ambeed
Warning

A179954-100G A179954-1G A179954-25G A179954-500G A179954-10G
77287-150EA 118 USD
77287-150 77287-152 77287-154 77287-156 77748-179
2-Amino-5-bromonicotinic acid 97% (by HPLC)
2-Amino-5-bromonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O₂
MDL Number: MFCD01860227
CAS Number: 52833-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR