(3-Glycidoxypropyl)triethoxysilane ≥98% (by GC)

Supplier: Ambeed
Warning

Synonyms: Geniosil GF 82, Glycidyl 3-Triethoxysilylpropyl ether, Triethoxy(3-glycidyloxypropyl)silane, (3-Glycidyloxypropyl)triethoxysilane, Triethoxy(3-(oxiran-2-ylmethoxy)propyl)silane, (3-(2,3-Epoxypropoxy)propyl)triethoxysilane

A179895-100G A179895-25G
77274-004EA 38 USD
77274-004 77274-006
(3-Glycidoxypropyl)triethoxysilane ≥98% (by GC)
(3-Glycidoxypropyl)triethoxysilane

Precaución: for Research Use Only

Formula: C₁₂H₂₆O₅Si
MDL Number: MFCD01311705
CAS Number: 2602-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR