(R)-(+)-2,2'-Bis[di(3,5-xylyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl 98%

Supplier: Ambeed
Warning

A179853-100MG A179853-1G A179853-250MG A179853-50MG
77248-234EA 145 USD
77248-234 77248-236 77248-238 77248-240
(R)-(+)-2,2'-Bis[di(3,5-xylyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl 98%
(R)-(+)-2,2'-Bis[di(3,5-xylyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₄₆H₄₈O₂P₂
MW: 694.8197 g/mol
MDL Number: MFCD09753009
CAS Number: 394248-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR