(R,E)-N-(4-(3-((5-Chloro-4-(1H-indol-3-yl)pyrimidin-2-yl)amino)piperidine-1-carbonyl)phenyl)-4-(dimethylamino)but-2-enamide 98%
Supplier: Ambeed
|
Warning
|
A179820-10MG
A179820-1MG
A179820-25MG
A179820-50MG
A179820-5MG
77273-994EA
140
USD
77273-994
77273-996
77273-998
77274-000
77274-002
(R,E)-N-(4-(3-((5-Chloro-4-(1H-indol-3-yl)pyrimidin-2-yl)amino)piperidine-1-carbonyl)phenyl)-4-(dimethylamino)but-2-enamide 98%
(R,E)-N-(4-(3-((5-Chloro-4-(1H-indol-3-yl)pyrimidin-2-yl)amino)piperidine-1-carbonyl)phenyl)-4-(dimethylamino)but-2-enamide
Precaución: for Research Use Only
Formula:
C₃₀H₃₂ClN₇O₂ |
MDL Number:
MFCD31563595 CAS Number: 1702809-17-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...