7-Methoxybenzofuran 97%

Supplier: Ambeed
Warning

A179818-1G A179818-250MG
77260-190EA 70 USD
77260-190 77260-192
7-Methoxybenzofuran 97%
7-Methoxybenzofuran

Precaución: for Research Use Only

Formula: C₉H₈O₂
MDL Number: MFCD01310825
CAS Number: 7168-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR