2-(6-Fluoro-1H-indol-3-yl)-N-(3-(2,2,3,3-tetrafluoropropoxy)benzyl)ethanamine hydrochloride 98+%

Supplier: Ambeed
Warning

Synonyms: Idalopirdine HCl

A179812-10MG A179812-1MG A179812-2MG A179812-50MG A179812-5MG
77236-522EA 22.66 USD
77236-522 77236-524 77236-526 77236-528 77236-530
2-(6-Fluoro-1H-indol-3-yl)-N-(3-(2,2,3,3-tetrafluoropropoxy)benzyl)ethanamine hydrochloride 98+%
2-(6-Fluoro-1H-indol-3-yl)-N-(3-(2,2,3,3-tetrafluoropropoxy)benzyl)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₂₀H₂₀ClF₅N₂O
MW: 434.83 g/mol
MDL Number: MFCD25372046
CAS Number: 467458-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR