4-Amino-3-ethylbenzonitrile 97%
Supplier: Ambeed
|
Danger
|
A179776-250MG
A179776-5G
A179776-1G
77248-232EA
11
USD
77248-232
77567-648
77746-171
4-Amino-3-ethylbenzonitrile 97%
4-Amino-3-ethylbenzonitrile
Precaución: For research use only.
Formula:
C₉H₁₀N₂ MW: 146.19 g/mol |
MDL Number:
MFCD00041431 CAS Number: 170230-87-2 UN: 3439 ADR: 6.1 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...