Propargyl-PEG3-Carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-(2-(2-(Prop-2-yn-1-yloxy)ethoxy)ethoxy)propanoic acid, Propargyl-PEG3-acid, 3-[2-[2-(2-Propyn-1-yloxy)ethoxy]ethoxy]propanoic acid, 3-[2-[2-(2-Propargyloxy)ethoxy]ethoxy]propanoic acid

A179722-100MG A179722-1G A179722-5G A179722-250MG
77260-180EA 28 USD
77260-180 77260-182 77748-177 77748-176
Propargyl-PEG3-Carboxylic acid 97%
Propargyl-PEG3-Carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₅
MW: 216.23 g/mol
MDL Number: MFCD22201521
CAS Number: 1347760-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR