3-Benzyloxy-2,6-difluorophenylboronic acid 98%

Supplier: Ambeed
Warning

A179707-1G A179707-250MG A179707-25G A179707-5G
77273-986EA 14 USD
77273-986 77273-988 77273-990 77273-992
3-Benzyloxy-2,6-difluorophenylboronic acid 98%
3-Benzyloxy-2,6-difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BF₂O₃
MDL Number: MFCD06200705
CAS Number: 870718-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR