(3R,4R,5R)-3,4-Bis(benzyloxy)-5-((benzyloxy)methyl)dihydrofuran-2(3H)-one 98%

Supplier: Ambeed
Warning

Synonyms: 2,3,5-Tri-O-benzyl-D-ribono-1,4-lactone

A179665-100G A179665-10G A179665-1G A179665-250MG A179665-25G A179665-5G
77236-506EA 307.97 USD
77236-506 77236-508 77236-510 77236-512 77236-514 77236-516
(3R,4R,5R)-3,4-Bis(benzyloxy)-5-((benzyloxy)methyl)dihydrofuran-2(3H)-one 98%
(3R,4R,5R)-3,4-Bis(benzyloxy)-5-((benzyloxy)methyl)dihydrofuran-2(3H)-one

Precaución: for Research Use Only

Formula: C₂₆H₂₆O₅
MW: 418.48 g/mol
MDL Number: MFCD08703966
CAS Number: 55094-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR