(2-Bromophenyl)acetone 97%

Supplier: Ambeed
Warning

A179632-1G A179632-5G A179632-25G
77260-172EA 17 USD
77260-172 77260-174 77748-174
(2-Bromophenyl)acetone 97%
(2-Bromophenyl)acetone

Precaución: for Research Use Only

Formula: C₉H₉BrO
MDL Number: MFCD03410431
CAS Number: 21906-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR