N-Boc-azetidine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: Boc-azetidine-3-carboxylic acid, N-Boc-Azetidine-3-carboxylic acid, 1-N-Boc-3-Azetidinecarboxylic acid, 1-Boc-azetidine-3-carboxylic Acid

A179533-100G A179533-10G A179533-1G A179533-25G A179533-5G A179533-500G
77248-222EA 118 USD
77248-222 77248-224 77248-226 77248-228 77248-230 77615-510
N-Boc-azetidine-3-carboxylic acid 97%
N-Boc-azetidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₁₅NO₄
MDL Number: MFCD01860897
CAS Number: 142253-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR