Methyl homovanillate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(4-hydroxy-3-methoxyphenyl)acetate

A179244-1G A179244-5G
77236-496EA 112.27 USD
77236-496 77236-498
Methyl homovanillate 98%
Methyl homovanillate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
MDL Number: MFCD00127727
CAS Number: 15964-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR